Assay Details
| Gene Symbol: BARX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204193.1 | NP_989524.1 | 1 | 151 | 103 | |
| GenBank mRNA | AB044371.1 | - | 1 | 151 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BARX1| Gene Symbol: BARX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204193.1 | NP_989524.1 | 1 | 151 | 103 | |
| GenBank mRNA | AB044371.1 | - | 1 | 151 | 103 | |
Set Membership: |
|