Assay Details
| Gene Symbol: CDKN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204433.1 | NP_989764.1 | 2 | 1657 | 91 | |
| GenBank mRNA | AY138247.1 | - | 2 | 1657 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDKN2B| Gene Symbol: CDKN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204433.1 | NP_989764.1 | 2 | 1657 | 91 | |
| GenBank mRNA | AY138247.1 | - | 2 | 1657 | 91 | |
Set Membership: |
|