Assay Details
| Gene Symbol: NDUFC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213574.1 | NP_998739.1 | 2 | 847 | 72 | |
| GenBank mRNA | AY568633.1 | - | 2 | 847 | 72 | |
| BX932511.1 | - | 2 | 822 | 72 | ||
| BX932719.1 | - | 2 | 845 | 72 | ||
| BX936000.1 | - | 2 | 822 | 72 | ||
| CR353099.1 | - | 3 | 464 | 72 | ||
More Information
Set Membership: |
|

NDUFC2
Genomic Map