Assay Details
| Gene Symbol: TARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318455.1 | NP_001305384.1 | 3 - 4 | 520 | 89 | |
| XM_025147355.1 | XP_025003123.1 | 3 - 4 | 1539 | 89 | ||
| GenBank mRNA | U22666.1 | - | 3 - 4 | 520 | 89 | |
| U78260.1 | - | 3 - 4 | 302 | 89 | ||
| U78284.1 | - | 2 - 3 | 544 | 89 | ||
More Information
Set Membership: |
|

TARP
Genomic Map