Assay Details
| Gene Symbol: TARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318455.1 | NP_001305384.1 | 5 - 6 | 902 | 113 | |
| XM_025147355.1 | XP_025003123.1 | 5 - 6 | 1921 | 113 | ||
| GenBank mRNA | U22666.1 | - | 5 - 6 | 902 | 113 | |
More Information
Set Membership: |
|

TARP
Genomic Map