Assay Details
Gene Symbol: UPF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318462.1 | NP_001305391.1 | 3 - 4 | 500 | 72 | |
GenBank mRNA | AJ721034.1 | - | 3 - 4 | 500 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
