Assay Details
| Gene Symbol: LYPLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162382.1 | NP_001155854.1 | 6 - 7 | 398 | 59 | |
| XM_015297672.2 | XP_015153158.1 | 12 - 13 | 1501 | 59 | ||
| XM_015297673.2 | XP_015153159.1 | 7 - 8 | 543 | 59 | ||
| GenBank mRNA | CD762320.1 | - | 7 - 8 | 494 | 59 | |
| DR410184.1 | - | 4 - 5 | 287 | 59 | ||
More Information
Set Membership: |
|

LYPLA2
Genomic Map