Assay Details
| Gene Symbol: MB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167752.2 | NP_001161224.1 | 2 - 3 | 376 | 89 | |
| GenBank mRNA | BU309726.1 | - | 2 - 3 | 376 | 89 | |
More Information
Set Membership: |
|

MB
Genomic Map