Assay Details
| Gene Symbol: SLC16A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321737.1 | NP_001308666.1 | 4 - 5 | 1088 | 63 | |
| GenBank mRNA | CR406820.1 | - | 2 - 3 | 406 | 63 | |
| KT876069.1 | - | 4 - 5 | 1088 | 63 | ||
More Information
Set Membership: |
|

SLC16A2
Genomic Map