Assay Details
| Gene Symbol: MTTPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322804.1 | NP_001309733.1 | 8 - 9 | 1069 | 104 | |
| XM_025151646.1 | XP_025007414.1 | 7 - 8 | 6092 | 104 | ||
| GenBank mRNA | KT899704.1 | - | 8 - 9 | 1069 | 104 | |
More Information
Set Membership: |
|

MTTPL
Genomic Map