Assay Details
| Gene Symbol: AvBD14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348511.1 | NP_001335440.1 | 2 - 3 | 300 | 107 | |
| XM_025148463.1 | XP_025004231.1 | 2 - 3 | 265 | 107 | ||
More Information
Additional Information:
				    	 
					    	
					        	
					            	This assay is in the 5'UTR region of the NM_001348511.1, XM_025148463.1 transcripts.
					        	
					    	            
				    	
			    	
			    
			Set Membership: | 
		   
					
						
							 | 
				

AvBD14
Genomic Map