Assay Details
| Gene Symbol: PRELID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204972.2 | NP_990303.2 | 3 - 4 | 524 | 76 | |
| GenBank mRNA | AJ397576.1 | - | 2 - 3 | 379 | 76 | |
| BX935862.1 | - | 3 - 4 | 369 | 76 | ||
| CR352501.1 | - | 3 - 4 | 524 | 76 | ||
More Information
Set Membership: |
|

PRELID1
Genomic Map