Assay Details
| Gene Symbol: BTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004769.1 | NP_001004769.1 | 2 - 3 | 172 | 93 | |
| XM_015276318.2 | XP_015131804.1 | 3 - 4 | 325 | 93 | ||
| XM_015276319.2 | XP_015131805.1 | 3 - 4 | 1677 | 93 | ||
| XM_015276320.2 | XP_015131806.1 | 2 - 3 | 417 | 93 | ||
| GenBank mRNA | AY681125.1 | - | 2 - 3 | 172 | 93 | |
| AY714349.1 | - | 2 - 3 | 172 | 93 | ||
More Information
Set Membership: |
|

BTC
Genomic Map