Assay Details
| Gene Symbol: TNFAIP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031441.2 | NP_001026612.1 | 1 - 2 | 184 | 92 | |
| GenBank mRNA | AJ720906.1 | - | 1 - 2 | 185 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ720906.1, NM_001031441.2 transcripts.
Set Membership: |
|

TNFAIP8
Genomic Map