Assay Details
| Gene Symbol: COX7A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198782.1 | NP_001185711.1 | 1 - 2 | 79 | 101 | |
| GenBank mRNA | BX257969.4 | - | 1 - 2 | 79 | 101 | |
| BX931842.1 | - | 1 - 2 | 71 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001198782.1 transcripts.
Set Membership: |
|

COX7A2
Genomic Map