Assay Details
| Gene Symbol: OST4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198884.1 | NP_001185813.1 | 2 - 3 | 180 | 71 | |
| GenBank mRNA | BX932399.1 | - | 1 - 2 | 153 | 71 | |
| BX934390.2 | - | 2 - 3 | 180 | 71 | ||
More Information
Set Membership: |
|

OST4
Genomic Map