Assay Details
Gene Symbol: NDUFV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199170.1 | NP_001186099.1 | 2 - 3 | 227 | 89 | |
XM_015300381.2 | XP_015155867.1 | 2 - 3 | 260 | 89 | ||
GenBank mRNA | DR428005.1 | - | 2 - 3 | 227 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
