Assay Details
| Gene Symbol: DNAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199681.1 | NP_001186610.1 | 1 - 2 | 155 | 66 | |
| XM_015287081.2 | XP_015142567.1 | 1 - 2 | 354 | 66 | ||
| XM_015287082.2 | XP_015142568.1 | 1 - 2 | 354 | 66 | ||
| XM_015287083.2 | XP_015142569.1 | 1 - 2 | 354 | 66 | ||
| XM_015287084.2 | XP_015142570.1 | 1 - 2 | 33 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001199681.1, XM_015287084.2 transcripts.
Set Membership: |
|

DNAL1
Genomic Map