Assay Details
| Gene Symbol: GATAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277498.1 | NP_001264427.1 | 4 - 5 | 651 | 82 | |
| GenBank mRNA | BU337426.1 | - | 3 - 4 | 297 | 82 | |
| BX934047.1 | - | 4 - 5 | 649 | 82 | ||
| DN928332.1 | - | 4 - 5 | 427 | 82 | ||
More Information
Set Membership: |
|

GATAD1
Genomic Map