Assay Details
| Gene Symbol: ZBTB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277639.2 | NP_001264568.1 | 2 - 3 | 112 | 108 | |
| XM_025156049.1 | XP_025011817.1 | 2 - 3 | 179 | 108 | ||
| GenBank mRNA | CR385386.1 | - | 2 - 3 | 112 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277639.2, XM_025156049.1 transcripts.
Set Membership: |
|

ZBTB26
Genomic Map