Assay Details
| Gene Symbol: NTMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277688.1 | NP_001264617.1 | 4 - 5 | 602 | 144 | |
| NM_001277689.1 | NP_001264618.1 | 4 - 5 | 563 | 144 | ||
| NM_001277690.1 | NP_001264619.1 | 4 - 5 | 570 | 144 | ||
| XM_015279519.2 | XP_015135005.1 | 4 - 5 | 700 | 144 | ||
| XM_015279521.2 | XP_015135007.1 | 4 - 5 | 738 | 144 | ||
| XM_015279522.2 | XP_015135008.1 | 4 - 5 | 664 | 144 | ||
| GenBank mRNA | BX934045.2 | - | 4 - 5 | 570 | 144 | |
More Information
Set Membership: |
|

NTMT1
Genomic Map