Assay Details
| Gene Symbol: NKX2-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277718.1 | NP_001264647.1 | 1 - 2 | 261 | 109 | |
| XM_015283379.2 | XP_015138865.1 | 2 - 3 | 936 | 109 | ||
| GenBank mRNA | AF110994.1 | - | 1 - 2 | 105 | 109 | |
More Information
Set Membership: |
|

NKX2-2
Genomic Map