Assay Details
| Gene Symbol: CCL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293309.1 | NP_001280238.1 | 1 - 2 | 123 | 117 | |
| GenBank mRNA | BI067703.1 | - | 1 - 2 | 108 | 117 | |
| DN829974.1 | - | 1 - 2 | 123 | 117 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001293309.1 transcripts.
Set Membership: |
|

CCL17
Genomic Map