Assay Details
| Gene Symbol: TEX264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293319.1 | NP_001280248.1 | 3 - 4 | 410 | 107 | |
| GenBank mRNA | BM488563.1 | - | 3 - 4 | 412 | 107 | |
More Information
Set Membership: |
|

TEX264
Genomic Map