Assay Details
| Gene Symbol: SEC61G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302188.1 | NP_001289117.1 | 2 - 3 | 140 | 97 | |
| XM_001236151.5 | XP_001236152.2 | 2 - 3 | 271 | 97 | ||
| XM_015281597.2 | XP_015137083.1 | 2 - 3 | 166 | 97 | ||
| GenBank mRNA | BU395054.1 | - | 2 - 3 | 140 | 97 | |
More Information
Set Membership: |
|

SEC61G
Genomic Map