Assay Details
| Gene Symbol: NAIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030692.2 | NP_001025863.2 | 2 | 2716 | 92 | |
| XM_015279495.2 | XP_015134981.1 | 3 | 2569 | 92 | ||
| XM_025156077.1 | XP_025011845.1 | 3 | 2589 | 92 | ||
| Gene Symbol: FAM102A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025156076.1 | XP_025011844.1 | 1 | 1450 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025156076.1 transcripts.
Set Membership: |
|

NAIF1
Genomic Map