Assay Details
| Gene Symbol: ZNF711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025150039.1 | XP_025005807.1 | 12 - 13 | 5520 | 75 | |
| GenBank mRNA | BX929449.1 | - | 4 - 5 | 858 | 76 | |
| BX931375.2 | - | 4 - 5 | 285 | 75 | ||
More Information
Set Membership: |
|

ZNF711
Genomic Map