Assay Details
| Gene Symbol: UBTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288795.1 | XP_015144281.1 | 3 | 103 | ||
| GenBank mRNA | BX929725.2 | - | 1 | 117 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBTD1| Gene Symbol: UBTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288795.1 | XP_015144281.1 | 3 | 103 | ||
| GenBank mRNA | BX929725.2 | - | 1 | 117 | 103 | |
Set Membership: |
|