Assay Details
| Gene Symbol: DDIT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288240.1 | XP_015143726.1 | 2 | 1327 | 147 | |
| GenBank mRNA | CR353438.1 | - | 1 | 822 | 147 | |
More Information
Set Membership: |
|

DDIT4
Genomic Map