Assay Details
| Gene Symbol: OSGIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015282839.2 | XP_015138325.2 | 4 - 5 | 902 | 86 | |
| XM_015282840.2 | XP_015138326.2 | 3 - 4 | 788 | 86 | ||
| XM_015282841.2 | XP_015138327.1 | 5 - 6 | 841 | 86 | ||
| XM_015282842.2 | XP_015138328.1 | 3 - 4 | 1142 | 86 | ||
| XM_015282843.2 | XP_015138329.1 | 3 - 4 | 473 | 86 | ||
| XM_025148031.1 | XP_025003799.1 | 4 - 5 | 441 | 86 | ||
| GenBank mRNA | CR353664.1 | - | 3 - 4 | 441 | 86 | |
More Information
Set Membership: |
|

OSGIN2
Genomic Map