Assay Details
| Gene Symbol: FAM109B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_425472.6 | XP_425472.2 | 2 | 3575 | 76 | |
| GenBank mRNA | CR387726.1 | - | 1 | 125 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM109B| Gene Symbol: FAM109B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_425472.6 | XP_425472.2 | 2 | 3575 | 76 | |
| GenBank mRNA | CR387726.1 | - | 1 | 125 | 76 | |
Set Membership: |
|