Assay Details
| Gene Symbol: ZCCHC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_424900.6 | XP_424900.1 | 5 | 137 | ||
| GenBank mRNA | CR389668.1 | - | 1 | 519 | 137 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZCCHC9| Gene Symbol: ZCCHC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_424900.6 | XP_424900.1 | 5 | 137 | ||
| GenBank mRNA | CR389668.1 | - | 1 | 519 | 137 | |
Set Membership: |
|