Assay Details
| Gene Symbol: BCL9L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298270.2 | XP_015153756.1 | 6 | 2653 | 100 | |
| XM_025143329.1 | XP_024999097.1 | 6 | 2756 | 100 | ||
| GenBank mRNA | CR390840.1 | - | 1 | 418 | 100 | |
More Information
Set Membership: |
|

BCL9L
Genomic Map