Assay Details
| Gene Symbol: LMCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414443.6 | XP_414443.4 | 6 | 4332 | 97 | |
| GenBank mRNA | CR406126.1 | - | 1 | 1119 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LMCD1| Gene Symbol: LMCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414443.6 | XP_414443.4 | 6 | 4332 | 97 | |
| GenBank mRNA | CR406126.1 | - | 1 | 1119 | 97 | |
Set Membership: |
|