Assay Details
| Gene Symbol: TMEM14C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015275973.2 | XP_015131459.1 | 1 - 2 | 331 | 92 | |
| GenBank mRNA | BX929916.2 | - | 1 - 2 | 324 | 92 | |
| BX930876.1 | - | 1 - 2 | 330 | 92 | ||
| BX930974.2 | - | 1 - 2 | 317 | 92 | ||
| BX932944.2 | - | 1 - 2 | 290 | 92 | ||
More Information
Set Membership: |
|

TMEM14C
Genomic Map