Assay Details
| Gene Symbol: VWA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004947404.3 | XP_004947461.1 | 6 | 3838 | 93 | |
| XM_417574.6 | XP_417574.3 | 6 | 3838 | 93 | ||
| GenBank mRNA | CR406450.1 | - | 1 | 1022 | 93 | |
More Information
Set Membership: |
|

VWA1
Genomic Map