Assay Details
| Gene Symbol: CCDC59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015283408.2 | XP_015138894.1 | 4 | 1917 | 101 | |
| GenBank mRNA | CR406952.1 | - | 1 | 119 | 101 | |
More Information
Set Membership: |
|

CCDC59
Genomic Map