Assay Details
| Gene Symbol: TMUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_428658.5 | XP_428658.5 | 1 - 2 | 507 | 80 | |
| GenBank mRNA | CR407023.1 | - | 1 - 2 | 73 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMUB1| Gene Symbol: TMUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_428658.5 | XP_428658.5 | 1 - 2 | 507 | 80 | |
| GenBank mRNA | CR407023.1 | - | 1 - 2 | 73 | 80 | |
Set Membership: |
|