Assay Details
| Gene Symbol: ARSJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420639.6 | XP_420639.6 | 1 - 2 | 860 | 74 | |
| GenBank mRNA | CR407275.1 | - | 1 - 2 | 152 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARSJ| Gene Symbol: ARSJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420639.6 | XP_420639.6 | 1 - 2 | 860 | 74 | |
| GenBank mRNA | CR407275.1 | - | 1 - 2 | 152 | 74 | |
Set Membership: |
|