Assay Details
| Gene Symbol: ZDHHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015281737.2 | XP_015137223.1 | 3 - 4 | 568 | 89 | |
| XM_015281738.2 | XP_015137224.1 | 3 - 4 | 505 | 89 | ||
| XM_015281741.2 | XP_015137227.1 | 3 - 4 | 568 | 89 | ||
| XM_015281742.2 | XP_015137228.1 | 3 - 4 | 568 | 89 | ||
| XM_025147503.1 | XP_025003271.1 | 3 - 4 | 505 | 89 | ||
| XM_025147504.1 | XP_025003272.1 | 3 - 4 | 546 | 89 | ||
| XM_025147505.1 | XP_025003273.1 | 3 - 4 | 478 | 89 | ||
| GenBank mRNA | CR407357.1 | - | 2 - 3 | 468 | 89 | |
More Information
Set Membership: |
|

ZDHHC3
Genomic Map