Assay Details
| Gene Symbol: ISM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015287724.2 | XP_015143210.1 | 6 | 4656 | 80 | |
| XM_015287725.2 | XP_015143211.1 | 6 | 4650 | 80 | ||
| GenBank mRNA | CR407426.1 | - | 1 | 1624 | 80 | |
More Information
Set Membership: |
|

ISM2
Genomic Map