Assay Details
| Gene Symbol: WDR34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415494.6 | XP_415494.2 | 5 - 6 | 708 | 97 | |
| GenBank mRNA | CR523255.1 | - | 2 - 3 | 237 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WDR34| Gene Symbol: WDR34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415494.6 | XP_415494.2 | 5 - 6 | 708 | 97 | |
| GenBank mRNA | CR523255.1 | - | 2 - 3 | 237 | 97 | |
Set Membership: |
|