Assay Details
| Gene Symbol: LCA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940421.3 | XP_004940478.1 | 8 | 3418 | 105 | |
| XM_419870.6 | XP_419870.1 | 8 | 3489 | 105 | ||
| GenBank mRNA | CR524234.1 | - | 1 | 615 | 105 | |
More Information
Set Membership: |
|

LCA5
Genomic Map