Assay Details
| Gene Symbol: HEBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422283.6 | XP_422283.5 | 5 | 2183 | 82 | |
| GenBank mRNA | CR524455.1 | - | 1 | 617 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HEBP2| Gene Symbol: HEBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422283.6 | XP_422283.5 | 5 | 2183 | 82 | |
| GenBank mRNA | CR524455.1 | - | 1 | 617 | 82 | |
Set Membership: |
|