Assay Details
| Gene Symbol: PPP2R2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945052.3 | XP_004945109.1 | 8 - 9 | 1066 | 54 | |
| XM_015294054.2 | XP_015149540.1 | 7 - 8 | 1101 | 54 | ||
| XM_414666.6 | XP_414666.3 | 7 - 8 | 1184 | 54 | ||
More Information
Set Membership: |
|

PPP2R2B
Genomic Map