Assay Details
| Gene Symbol: GLDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015291961.2 | XP_015147447.1 | 9 - 10 | 1107 | 72 | |
| XM_025154023.1 | XP_025009791.1 | 9 - 10 | 825 | 72 | ||
More Information
Set Membership: |
|

GLDN
Genomic Map