Assay Details
| Gene Symbol: FBXO28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015283908.2 | XP_015139394.1 | 2 - 3 | 366 | 102 | |
| XM_015283909.2 | XP_015139395.1 | 2 - 3 | 371 | 102 | ||
More Information
Set Membership: |
|

FBXO28
Genomic Map