Assay Details
| Gene Symbol: TTBK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004941660.3 | XP_004941717.1 | 15 - 16 | 3969 | 72 | |
| XM_015287100.2 | XP_015142586.1 | 15 - 16 | 3969 | 72 | ||
| XM_015287101.2 | XP_015142587.1 | 10 - 11 | 3093 | 72 | ||
| XM_015287102.2 | XP_015142588.1 | 10 - 11 | 3065 | 72 | ||
| XM_421164.6 | XP_421164.3 | 14 - 15 | 3720 | 72 | ||
More Information
Set Membership: |
|

TTBK2
Genomic Map