Assay Details
| Gene Symbol: GID4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414816.5 | XP_414816.2 | 5 - 6 | 759 | 87 | |
| GenBank mRNA | BX930655.2 | - | 5 - 6 | 700 | 87 | |
| BX936115.1 | - | 5 - 6 | 664 | 87 | ||
More Information
Set Membership: |
|

GID4
Genomic Map