Assay Details
| Gene Symbol: GPR63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003641015.4 | XP_003641063.1 | 2 | 4957 | 152 | |
| XM_015284676.2 | XP_015140162.1 | 3 | 5073 | 152 | ||
More Information
Set Membership: |
|

GPR63
Genomic Map